Skip to Content
Merck
All Photos(1)

Documents

738646

Sigma-Aldrich

trans-2-Heptene

99%

Synonym(s):

(2E)-2-Heptene, (E)-2-Heptene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CHCH3
CAS Number:
Molecular Weight:
98.19
Beilstein:
1719390
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

88 mmHg ( 37.7 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.404 (lit.)
n20/D 1.405

bp

98 °C (lit.)

density

0.701 g/mL at 25 °C (lit.)
0.705 g/mL at 25 °C

SMILES string

CCCC\C=C\C

InChI

1S/C7H14/c1-3-5-7-6-4-2/h3,5H,4,6-7H2,1-2H3/b5-3+

InChI key

OTTZHAVKAVGASB-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

26.1 °F

Flash Point(C)

-3.3 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyuki Ohno et al.
Journal of AOAC International, 97(5), 1452-1458 (2015-04-24)
A headspace GC/MS method was developed for identification and quantitation of residual volatile organic compounds in poly(methyl methacrylate) (PMMA) kitchen utensils. A sample was cut into small pieces, then N,N-dimethylacetamide was added in a headspace vial and sealed. After storing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service