Skip to Content
Merck
All Photos(1)

Documents

436712

Supelco

Amberlyst® 36 Ion Exchange Resin

16-50 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

product name

Amberlyst® 36, total pore volume 0.20 mL/g

form

beads

Quality Level

crosslinking

12 % cross-linked

contains

51-57% Moisture content (H+ form)

composition

water, 55%

technique(s)

LPLC: suitable

surface area

33 m2/g

matrix

styrene divinylbenzene (macroporous)

matrix active group

sulfonic acid functional group

particle size

<0.425 mm (0.5% max)
0.600-0.850 mm (uniformity coefficient: <1.6)
>1.180 mm (4.0% max)

pore size

0.20 mL/g total pore volume
240 Å average pore diameter

capacity

>1.95 eq/L exchange capacity
>5.40 meq/g exchange capacity

density

1.2 g/mL at 25 °C (lit.)

separation technique

cation exchange

InChI

1S/C10H10.C8H8O3S/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-5-3-4-6-8(7)12(9,10)11/h3-8H,1-2H2;2-6H,1H2,(H,9,10,11)

InChI key

SIWVGXQOXWGJCI-UHFFFAOYSA-N

General description

Shrinkage:
Water to phenol: 45%
Water to dry: 54%

Application


  • Synthesis of renewable diesel and jet fuel range alkanes using 2-methylfuran and cyclohexanone.: This study explores a method for producing renewable diesel and jet fuel from biomass-derived compounds using Amberlyst® 36 as a catalyst, highlighting its potential for sustainable fuel production (Sun et al., 2022).

  • Kinetic Study on Esterification of Acetic Acid with Isopropyl Alcohol Catalyzed by Ion Exchange Resin.: The research investigates the kinetics of esterification reactions catalyzed by Amberlyst® 36, providing insights into its efficiency and applicability in producing esters used in various industrial applications (Liu et al., 2019).

  • Pretreatment of bio-oil with ion exchange resin to improve fuel quality and reduce char during hydrodeoxygenation upgrading with Pt/C.: This paper discusses the use of Amberlyst® 36 in the pretreatment of bio-oil to enhance its quality as a precursor for sustainable fuel, demonstrating the resin′s effectiveness in removing impurities that affect fuel refinement processes (Oh et al., 2021).

  • Hydrolysis of Lipid-Extracted Chlorella vulgaris by Simultaneous Use of Solid and Liquid Acids.: The study examines the hydrolysis of algae-derived lipids using a combination of Amberlyst® 36 and other acids, showcasing its potential to facilitate biofuel production from microalgae (Seon et al., 2019).

  • Valorization of lignocellulosic fibres of paper waste into levulinic acid using solid and aqueous Brønsted acid.: This research demonstrates the role of Amberlyst® 36 in converting paper waste into valuable chemicals like levulinic acid, contributing to waste valorization and chemical recycling (Chen et al., 2018).

Legal Information

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service