Skip to Content
Merck
All Photos(2)

Documents

416738

Sigma-Aldrich

S-tert-Butyl thioacetate

98%

Synonym(s):

1-(tert-Butylsulfanyl)ethan-1-one, S-(1,1-Dimethylethyl) ethanethioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COSC(CH3)3
CAS Number:
Molecular Weight:
132.22
Beilstein:
1740436
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

135-136 °C (lit.)

density

0.927 g/mL at 25 °C (lit.)

SMILES string

CC(=O)SC(C)(C)C

InChI

1S/C6H12OS/c1-5(7)8-6(2,3)4/h1-4H3

InChI key

PJFHXBBYLAVSLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

S-tert-Butyl thioacetate is a thioester.

Application

S-tert-Butyl thioacetate may be used for the preparation of vinyloxyborane derivative.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic studies on an antitumor antibiotic, bleomycin. XIV. The synthesis of Boc-pyrimidoblamic acid.
M Otsuka et al.
Chemical & pharmaceutical bulletin, 33(2), 520-526 (1985-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service