Skip to Content
Merck
All Photos(2)

Documents

371335

Sigma-Aldrich

4′-Hydroxy-4-biphenylcarboxylic acid

99%

Synonym(s):

4-(4-Hydroxyphenyl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4C6H4CO2H
CAS Number:
Molecular Weight:
214.22
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

99%

form

powder, crystals or chunks

mp

295 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC(=O)c1ccc(cc1)-c2ccc(O)cc2

InChI

1S/C13H10O3/c14-12-7-5-10(6-8-12)9-1-3-11(4-2-9)13(15)16/h1-8,14H,(H,15,16)

InChI key

JTGCXYYDAVPSFD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service