Skip to Content
Merck
All Photos(1)

Key Documents

279641

Sigma-Aldrich

Sodium tetrakis(1-imidazolyl)borate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12BN8Na
CAS Number:
Molecular Weight:
302.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

SMILES string

[Na+].c1cn(cn1)[B-](n2ccnc2)(n3ccnc3)n4ccnc4

InChI

1S/C12H12BN8.Na/c1-5-18(9-14-1)13(19-6-2-15-10-19,20-7-3-16-11-20)21-8-4-17-12-21;/h1-12H;/q-1;+1

InChI key

OVRFSDYWKKOQMW-UHFFFAOYSA-N

Application

Sodium tetrakis(1-imidazolyl)borate was used in the:
  • synthesis of series of zwitterionic molecules
  • preparation of poly(imidazolyl)borate organotin(IV) complex
  • precipitation of deuterium cations

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Study of Sodium Tetrakis (1-imidazolyl) borate in the Precipitation of Deuterium Cations.
Hockman DC, et al.
Analytical Letters, 14(9), 719-730 (1981)
Huaping Li et al.
Journal of the American Chemical Society, 131(25), 8903-8912 (2009-05-23)
A series of zwitterionic molecules were synthesized by addition of iodoalkanes to sodium tetrakis(1-imidazolyl)borate (NaBIm(4)). Single substitution at the N3 site leads to compounds designated as C(n)-BIm(4) (n = 1, 6, 12, 16), where the subscript corresponds to the number
Synthesis and characterization of the first poly (imidazolyl) borate organotin (IV) complex exhibiting a polymeric chain structure.
Pellei M, et al.
Inorganic Chemistry Communications, 4(12), 708-711 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service