Merck
  • Quaternary ammonium N,N,N-trimethyl chitosan derivative and povidone‑iodine complex as a potent antiseptic with enhanced wound healing property.
Quaternary ammonium N,N,N-trimethyl chitosan derivative and povidone‑iodine complex as a potent antiseptic with enhanced wound healing property.
International journal of biological macromolecules (2021-05-30)
Celine Abueva, Hyun Seok Ryu, Jun Won Min, Phil Sang Chung, Hyeon Sook You, Myung Suk Yang, Seung Hoon Woo
ABSTRACT
The importance of developing more potent antimicrobials and robust infection prevention practices has been highlighted recently with the increase in reports of emerging bacterial resistance mechanisms and the development of antibiotic-resistant microbes. In this study, a quaternary ammonium chitosan derivative, N,N,N-trimethyl chitosan chloride (TMC) with inherent bactericidal property was synthesized and complexed with povidone‑iodine (PVP-I) to create a potentially more potent antiseptic solution that could also significantly enhance the wound healing process. TMC, a positively charged, water-soluble derivative of chitosan, formed stable solutions with PVP-I at 5% w/v TMC concentration (TMC5/PVP-I). TMC5/PVP-I was significantly effective against multidrug-resistant bacteria S. aureus compared with PVP-I alone. TMC/PVP-I solutions also showed fungicidal property against C. albicans, with no cytotoxic effects when tested against human fibroblast cells cultured in vitro. Wound healing assessment in vivo revealed early collagen formation and re-epithelialization for TMC5/PVP-I treated wounds in rats relative to control and PVP-I only. Formulation of TMC/PVP-I solutions presented in the study can be easily adapted in the existing production of commercial PVP-I creating a new product with more potent bactericidal and enhanced wound healing properties for optimal wound care.
Sigma-Aldrich
LB Broth (Miller), Liquid microbial growth medium