Select a Size
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
triptorelin
form
solid
manufacturer/tradename
USP
application(s)
USP Biologics
pharmaceutical (small molecule)
format
neat
storage temp.
−20°C
General description
Peptide family: Triptorelin
This compound is associated with GnRH analogs utilized in hormone-sensitive conditions.
The USP biologics peptides category encompasses a diverse range of therapeutic peptides that are essential in managing various medical conditions. These peptides, typically consisting of amino acid sequences of 40 residues or less, are critical for the development of high-quality medicines. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of peptide therapeutics throughout their lifecycle.
The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.
Analysis Note
Other Notes
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.