Z740332
PierceASeal Foil™ heat sealable seal
LabRoll™, W × L 78 mm × 610 m, sterile
Synonym(s):
Pierceable foil seal, reusable heat seal
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Recommended Products
sterility
sterile
feature
piercable
packaging
pack of 1 ea
manufacturer/tradename
IST Scientific IST-106-078SR
W × L
78 mm × 610 m
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Pierceable foil seal with good solvent resistance.
Features: Foil, high solvent resistance including DMSO, non-peelable, multiple sealing/re-sealing permissible, easy sealing surface identification, pierceable.
Compatibility: Polypropylene (PP), Polystyrene (PS).
Seal integrity range: -20°C to 120°C.
Features: Foil, high solvent resistance including DMSO, non-peelable, multiple sealing/re-sealing permissible, easy sealing surface identification, pierceable.
Compatibility: Polypropylene (PP), Polystyrene (PS).
Seal integrity range: -20°C to 120°C.
Application
PCR, compound storage,sample shipping, can be sealedover existing seal
Legal Information
LabRoll is a trademark of IST Scientific
PierceASeal Foil is a trademark of IST Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service