TMO332189-0002
Nalgene® economy wide-mouth bottle
natural high-density polyethylene, volume 60 mL, case of 1000 ea
Synonym(s):
HDPE bottle, nalgene HDPE bottle
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
natural high-density polyethylene
packaging
case of 1000 ea
manufacturer/tradename
Nalgene 332189-0002
capacity
60 mL
volume
60 mL
Related Categories
General description
Perform short-term general purpose laboratory and field applications with Thermo Scientific Nalgene Wide-Mouth HDPE Economy Bottles with Closure. These light-duty translucent bottles are a lightweight version of the original Nalgene bottle, bulk-packed for ultimate economy. Thinner-walled and more flexible than the Nalgene Lab Quality Bottles, they feature a wide mouth for easy filling and offer excellent chemical resistance to most acids, bases and alcohols.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)