Skip to Content
Merck

SML3359

TP-040

≥98% (HPLC)

Synonym(s):

N-((1-Methyl-1H-imidazol-4-yl)methyl)-4-(4-methylpiperidin-1-yl)pyrimidin-2-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H22N6
CAS Number:
Molecular Weight:
286.38
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(CC1)CCN1C2=CC=NC(NCC3=CN(C)C=N3)=N2

InChI

1S/C15H22N6/c1-12-4-7-21(8-5-12)14-3-6-16-15(19-14)17-9-13-10-20(2)11-18-13/h3,6,10-12H,4-5,7-9H2,1-2H3,(H,16,17,19)

InChI key

PWKAYICUBVNJAZ-UHFFFAOYSA-N

Biochem/physiol Actions

Brain penetrant, potent and selective inhibitor of O-GlcNAcase (OGA).
TP-040 is a brain penetrant, potent and selective inhibitor of O-GlcNAcase (OGA). TP-040 increases the level of O-GlcNAcylated protein in cells.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library