Skip to Content
Merck

SML2559

C-176

≥98% (HPLC)

Synonym(s):

C 176, C176, N-(4-Iodophenyl)-5-nitro-2-furancarboxamide, N-(4-Iodophenyl)-5-nitrofuran-2-carboxamide, Sting inhibitor 1, Sting inhibitor C-176

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H7IN2O4
CAS Number:
Molecular Weight:
358.09
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to very dark gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(I)C=C1)C2=CC=C([N+]([O-])=O)O2

InChI

1S/C11H7IN2O4/c12-7-1-3-8(4-2-7)13-11(15)9-5-6-10(18-9)14(16)17/h1-6H,(H,13,15)

InChI key

JBIKQXOZLBLMKI-UHFFFAOYSA-N

Biochem/physiol Actions

C-176 is a potent and selective inhibitor against murine, but not human, stimulator of interferon genes (STING) via covalent modifiation of mSTING Cys91. C-176 inhibits STING-mediated IFNβ reporter activity (IC50 <50 nM) without affecting RIG-I, and blocks (1 μM) downstream TBK1 phosphorylation induction by STING agonist CMA (1.4 h) in mSTING-expressing HEK293T cells. C-176 markedly reduces CMA-induced serum IL-6 & type I IFNs in mice (by ≥85% 4 h post CMA; 750 nmol C-176/mouse i.p. 1 or 4 h prior to 224 mg/kg CMA) and significantly ameliorates the autoinflammatory severity in Trex1−/− mice.
Potent and selective, Cys91-targeting covalent inhibitor against murine, but not human, STING in vitro and in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library