Skip to Content
Merck
All Photos(1)

Key Documents

SML1261

Sigma-Aldrich

Asimadoline hydrochloride

≥98% (HPLC)

Synonym(s):

EMD-61753 hydrochloride, N-[(1S)-2-[(3S)-3-hydroxy-1-pyrrolidinyl]-1-phenylethyl]-N-methyl-a-phenyl-benzeneacetamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30N2O2 · HCl
CAS Number:
Molecular Weight:
451.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN([C@H](CN1C[C@@H](O)CC1)C2=CC=CC=C2)C(C(C3=CC=CC=C3)C4=CC=CC=C4)=O.Cl

InChI

1S/C27H30N2O2.ClH/c1-28(25(21-11-5-2-6-12-21)20-29-18-17-24(30)19-29)27(31)26(22-13-7-3-8-14-22)23-15-9-4-10-16-23;/h2-16,24-26,30H,17-20H2,1H3;1H/t24-,25+;/m0./s1

InChI key

GMJSLABTEURMBF-CLSOAGJSSA-N

Biochem/physiol Actions

Asimadoline is a potent and selective κ-opioid agonist that does not penetrate the blood-brain barrier. Asimadoline produces both analgesic and antidiarrheal effects.
Asimadoline is a potent and selective κ-opioid agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service