Skip to Content
Merck
All Photos(1)

Key Documents

SMB00087

Sigma-Aldrich

Visnadine

≥95% (LC/MS-ELSD)

Synonym(s):

Cardine, Carduben, Carduben-S, Selva, Visnadin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O7
CAS Number:
Molecular Weight:
388.41
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC[C@@H](C)C(=O)O[C@@H]1[C@H](OC(C)=O)c2c(OC1(C)C)ccc3C=CC(=O)Oc23

InChI

1S/C21H24O7/c1-6-11(2)20(24)27-19-18(25-12(3)22)16-14(28-21(19,4)5)9-7-13-8-10-15(23)26-17(13)16/h7-11,18-19H,6H2,1-5H3/t11-,18-,19-/m1/s1

InChI key

GVBNSPFBYXGREE-CXWAGAITSA-N

General description

Natural product derived from plant source.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit Regev Krugwasser et al.
Journal of vision, 19(4), 14-14 (2019-04-06)
The sense of agency (SoA) is the sensation of control over our actions. SoA is thought to rely mainly upon the comparison of predictions regarding the sensory outcomes of one's actions and the actual sensory outcomes. Previous studies have shown
J Durate et al.
Planta medica, 63(3), 233-236 (1997-06-01)
Visnadine, an active principle extracted from the fruit of Ammi visnaga, exhibits peripheral and coronary vasodilator activities and has been used for the treatment of angina pectoris. The present study was undertaken to further characterize the inhibitory effects of visnadine
G Zgórka et al.
Journal of chromatography. A, 797(1-2), 305-309 (1998-05-16)
A new, simple and rapid solid-phase extraction method for the determination of furanochromones and pyranocoumarins in Ammi visnaga L. fruits and pharmaceuticals by reversed-phase high-performance liquid chromatography (RP-HPLC) was developed. The isolation of compounds examined was carried out on octadecyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service