Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502185

Sigma-Aldrich

Anti-Progesterone Receptor antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NR3C3, PGR, PR, PRGR, nuclear receptor 3C3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 98 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PGR(5241)

General description

Anti-Progesterone Receptor Antibody detects endogenous levels of total Progesterone Receptor protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Progesterone Receptor.

Immunogen Range: 371-420

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yosuke Sugino et al.
The Journal of reproduction and development, 68(4), 254-261 (2022-06-02)
In ruminants, uterine glands play key roles in the establishment of pregnancy by secreting various factors into the uterine lumen. Although a three-dimensional (3D) culture system has been used for investigating cellular functions in vitro, the detailed functions of uterine
Yoshihiko Kobayashi et al.
Reproduction (Cambridge, England), 151(6), 577-587 (2016-03-05)
Nitric oxide (NO) is a regulator of sperm motility, oocyte/embryo survival, and waves of contraction/relaxation in mammalian oviducts. As follicles control oviductal functions by two routes at least, (1) a systemic way via blood vessels before ovulation, (2) a direct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service