Skip to Content

P2664

Pyroglutamic acid p-nitroanilide

protease substrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H11N3O4
CAS Number:
Molecular Weight:
249.22
NACRES:
NA.28
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (TLC)

form

powder

solubility

DMF: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

O=C1CC[C@H](N1)C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C11H11N3O4/c15-10-6-5-9(13-10)11(16)12-7-1-3-8(4-2-7)14(17)18/h1-4,9H,5-6H2,(H,12,16)(H,13,15)/t9-/m0/s1

InChI key

HGNBEWLBSCSJGV-VIFPVBQESA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library