Skip to Content
Merck

Skip To

MSQC26

SILuLite SigmaMAb Nivolumab Monoclonal Antibody

recombinant, expressed in CHO cells

Synonym(s):

Biologic Antibody Mimic, Biotherapeutic Antibody Mimic, Monoclonal Antibody Calibrator

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
12352203
Clone:
-
Species reactivity:
-
Application:
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in CHO cells

Quality Segment

assay

≥90% (HPLC)

form

solid

suitability

suitable for mass spectrometry

shipped in

wet ice

storage temp.

−20°C

General description

SigmaMAb Nivolumab (MSQC26) was designed to be a standard for the analysis of Nivolumab monoclonal antibodies using mass spectrometry. An isotopically labeled version is available as SILuMab Nivolumab (MSQC27).

SigmaMAb Nivolumab is for research use only and is not intended for diagnostic or therapeutic use.

Other Notes

SigmaMAb Nivolumab Heavy Chain:
QVQLVESGGGVVQPGRSLRLDCKASGITFSNSGMHWVRQAPGKGLEWVAVIWYDGSKRYYADSVKGRFTISRDNSKNTLFLQMNSLRAEDTAVYYCATNDDYWGQGTLVTVSSASTKGPSVFPLAPCSRSTSESTAALGCLVKDYFPEPVTVSWNSGALTSGVHTFPAVLQSSGLYSLSSVVTVPSSSLGTKTYTCNVDHKPSNTKVDKRVESKYGPPCPPCPAPEFLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSQEDPEVQFNWYVDGVEVHNAKTKPREEQFNSTYRVVSVLTVLHQDWLNGKEYKCKVSNKGLPSSIEKTISKAKGQPREPQVYTLPPSQEEMTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSRLTVDKSRWQEGNVFSCSVMHEALHNHYTQKSLSLSLG

SigmaMAb Nivolumab Light Chain:
EIVLTQSPATLSLSPGERATLSCRASQSVSSYLAWYQQKPGQAPRLLIYDASNRATGIPARFSGSGSGTDFTLTISSLEPEDFAVYYCQQSSNWPRTFGQGTKVEIKRTVAAPSVFIFPPSDEQLKSGTASVVCLLNNFYPREAKVQWKVDNALQSGNSQESVTEQDSKDSTYSLSSTLTLSKADYEKHKVYACEVTHQGLSSPVTKSFNRGEC

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSQC28MSQC27MSQC14
storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

solid

form

solid

form

solid

form

solid

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

assay

≥90% (HPLC)

assay

≥90% (HPLC)

assay

≥90% (HPLC)

assay

≥90% (SDS-PAGE)

recombinant

expressed in CHO cells

recombinant

expressed in CHO cells

recombinant

expressed in CHO cells

recombinant

expressed in CHO cells

suitability

suitable for mass spectrometry

suitability

suitable for mass spectrometry

suitability

suitable for mass spectrometry

suitability

suitable for mass spectrometry


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters