Merck
HS1419B
Pour Boat Weighing Dishes
medium, white polystyrene, pack of 500 ea
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.71
UNSPSC Code:
41121812
Material:
white polystyrene
Packaging:
pack of 500 ea
Manufacturer/tradename:
Heathrow Scientific HS1419B
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
white polystyrene
packaging
pack of 500 ea
manufacturer/tradename
Heathrow Scientific HS1419B
L × W × H
83 mm × 132 mm × 26 mm
General description
Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
Features and Benefits
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • pour-spout design easily dispenses samples without flexing the dish
  • flat-bottom dishes are made from 12 mil think polystyrene for better strength, stability, and handling
Still not finding the right product?
Explore all of our products under Pour Boat Weighing Dishes
Choose from one of the most recent versions:
Certificates of Analysis (COA)