Skip to Content
Merck
All Photos(8)

Key Documents

HPA001193

Sigma-Aldrich

ANTI-MAGEB1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CT3.1, Anti-Cancer/testis antigen 3.1, Anti-DAM10, Anti-DSS-AHC critical interval MAGE superfamily 10, Anti-MAGE-B1 antigen, Anti-MAGE-XP antigen, Anti-MAGEB1, Anti-MAGEB4, Anti-Melanoma-associated antigen B1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

FMNVLGAYDGEEHLIYGEPRKFITQDLVQEKYLKYEQVPNSDPPRYQFLWGPRAYAETTKMKVLEFLAKMNGATPRDFPSHYEEALRDEEERAQVRSSVRARRRTTATTFRARSRAPFSRSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAGEB1(4112)

Looking for similar products? Visit Product Comparison Guide

Specificity

Note: The Ensemble Gene ID has changed from ENSG00000120289 in the 46:36 release of the database to ENSG00000214107 in the 48:36 release of the database.

Immunogen

Melanoma-associated antigen B1 recombinant protein epitope signature tag (PrEST)

Application

Anti-MAGEB4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

MAGEB1 (Melanoma-associated antigen B1) gene belongs to the MAGEB family of genes. It is localized in the DSS (dosage-sensitive sex reversal) critical region. It is expressed in testis and tumors. It is located on chromosome Xp22-p21 along with other members of the family.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74072

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Dabovic et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 6(9), 571-580 (1995-09-01)
Patients with an intact SRY gene and duplications of portions of Xp21 develop as phenotypic females. We have recently mapped this sex reversal locus, DSS, to a 160-kb region of Xp21 that includes the adrenal hypoplasia congenita locus. To clone
F Muscatelli et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(11), 4987-4991 (1995-05-23)
A human gene with strong homology to the MAGE gene family located in Xq27-qter has been isolated by using exon-trapping of cosmids in the Xp21.3 region. We have mapped and sequenced cDNA and genomic clones corresponding to this gene, MAGE-Xp
Estelle Lecluze et al.
Human reproduction (Oxford, England), 35(5), 1099-1119 (2020-05-16)
Which transcriptional program triggers sex differentiation in bipotential gonads and downstream cellular events governing fetal testis and ovary development in humans? The characterization of a dynamically regulated protein-coding and non-coding transcriptional landscape in developing human gonads of both sexes highlights

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service