Key Documents
G6778
Sigma-Aldrich
Gaskets for vertical standard gel
16.5 cm wide, size 1.0 mm × 19 cm, color-coded red
About This Item
UNSPSC Code:
41105305
NACRES:
NB.51
Product Name
Gaskets for vertical standard gels, 16.5 cm wide, size 1.0 mm × 19 cm, color-coded red
size
1.0 mm × 19 cm
color
color-coded red
Features and Benefits
Unique silicone rubber gasket provides continuous seal around sides and bottom of glass plates, eliminating the need for a bottom spacer. Order gasket thickness to match comb and spacers. Gaskets are color-coded by thickness for easy identification. Length given is height of assembly; actual gasket length is 2 × h + w.
Choose from one of the most recent versions:
Certificates of Analysis (COA)