Skip to Content
Merck

D8687

Dispo-Biodialyzer

MWCO 1 kDa

Synonym(s):

Harvard Bioscience 740702, dialysis unit, dialyzer, disposable dialysis unit, small sample diallysis unit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41123100
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 50 ea

technique(s)

analytical sample preparation: suitable

pore size

1 kDa MWCO

application(s)

life science and biopharma

General description

A unique disposable unit for small sample dialysis, 5-100 μL. Dialysis is rapid with the ultra thin membrane of regenerated cellulose. After dialysis, sample can be recovered, almost 100% by centrifugation in a microcentrifuge tube. Recommended for salt removal, buffer exchange, protein binding assays and as a equilibrium dialyzer.

The Micro DispoDIALYZER includes a membrane, foam float, cap and two collection tubes (1.5 mL).


Still not finding the right product?

Explore all of our products under Dispo-Biodialyzer




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library