Skip to Content
Merck

CPS1A60

Chemiluminescent Peroxidase Substrate-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.83
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

usage

sufficient for 60 mL substrate

storage temp.

2-8°C

Application

Chemiluminescent Peroxidase Substrate can be used for the highly sensitive detection of peroxidase labeled material in a variety of Western blotting applications. This substrate is an enhanced luminol product with a stabilized peroxide buffer solution that provides femtogram sensitivity with minimal background interference.


Still not finding the right product?


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library