CLS354221
Corning® rLaminin-521
from human, pack of 100 μg
Synonym(s):
extracellular matrix
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
packaging
pack of 100 μg
manufacturer/tradename
Corning 354221
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning rLaminin-521, Human, 100μg Vial Corning rLaminin-521 (Human) is a heterotrimer composed of a5, ß2, and ?1 chains expressed in a mammalian cell culture system. rLaminin-521 (Human) supports long-term self-renewal of human pluripotent stem cells (hPSC), including embryonic stem cells (hESC) and induced pluripotent stem cells (iPSC) in defined and xeno-free environments. rLaminin-521 provides additional benefits, including ROCK inhibitor independent single cell expansion of PSCs, which improves hPSC culture ease and efficiency.
Features
Recombinant protein
Easy to self-coat
Available in 0.1 mg aliquots for easy handling
Compatible with multiple hPSC culture media and dissociation reagents
Supports undifferentiated expansion of hPSC
Suitable for xeno-free, defined culture environment
Features
Recombinant protein
Easy to self-coat
Available in 0.1 mg aliquots for easy handling
Compatible with multiple hPSC culture media and dissociation reagents
Supports undifferentiated expansion of hPSC
Suitable for xeno-free, defined culture environment
Legal Information
Corning is a registered trademark of Corning, Inc.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service