Skip to Content
Merck

DMS 454

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 95062832, human lung, Polygonal

Synonym(s):

DMS454 Cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
human lung
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
Not specified
Morphology:
Polygonal
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human lung

packaging

tube of 5 μg 95062832-DNA-5UG, pkg of vial of cells 95062832-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Polygonal

products

Calcitonin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

Human lung tumour model, hormone secretion

Biochem/physiol Actions

Human Caucasian lung, small cell carcinoma
The small cell carcinoma DMS 454 was isolated in 1981 from lung tissue of a male patient at time of autopsy. The cells contain dense-core vesicles and secrete calcitonin.

Preparation Note

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Waymouth′s MB 752/1 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (heat-inactivated).

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to [email protected].




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library