Skip to Content
Merck

KYSE-70

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 94072012, human esophagus, Epithelial

Synonym(s):

KY70 Cells, KYSE70 Cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
Biological source:
human esophagus
Relevant disease(s):
cancer
Growth mode:
Adherent
Karyotype:
Hypotriploid with 8% polyploidy
Morphology:
Epithelial
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human esophagus

packaging

tube of 5 μg 94072012-DNA-5UG, pkg of vial of cells 94072012-1VL

growth mode

Adherent

karyotype

Hypotriploid with 8% polyploidy

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Biochem/physiol Actions

Established from a poorly differentiated invasive oesophageal squamous cell carcinoma resected from middle intra-thoracic oesophagus of a 77-year-old Japanese man prior to treatment. This cell line has been mycoplasma eradicated at ECACC. By short tandem repeat (STR)-PCR analysis the Y chromosome could not be detected in this cell line. It is a known phenomenon that SSC cell lines can lose their Y chromosome.
Human oesophageal squamous cell carcinoma

Preparation Note

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)
Split confluent cultures 1:5 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Doubling time ~35 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library