Skip to Content
Merck

C2692

Nalgene® round carboy, with handles

nominal capacity 10 L (2.5 gal), low-density polyethylene

Synonym(s):

carboys, nalgene carboys, Nalgene 2210-0020, laboratory container, receptical, vessel

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41121820
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

low-density polyethylene , white polypropylene screw closure

packaging

pack of 1 ea

manufacturer/tradename

Nalgene 2210-0020

H × D

15 3/8 in. (389 mm) × 9 7/8 in. (249 mm)

nominal capacity

10 L (2.5 gal)

General description

Graduated in 1 gal and 5 L increments. 83B (83 mm) closure.
Store and transport reagents with in round LDPE Carboys with screw cap, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library