Skip to Content
Merck

58901C

Lactalbumin Hydrolysate Solution (50X)

liquid, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Lactalbumin enzymatic hydrolysate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

For research or for further manufacturing use

Quality Segment

sterility

sterile; sterile-filtered

form

liquid

technique(s)

cell culture | insect: suitable, cell culture | mammalian: suitable

shipped in

ambient

storage temp.

2-8°C

Application

Prepared with 166.5 g/L of lactalbumin enzymatic hydrolysate (LAH) in cell culture grade water. LAH is specifically formulated for use with insect cell culture media.


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library