Skip to Content
Merck
All Photos(1)

Documents

342R-1

Sigma-Aldrich

Tryptase (EP259) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP259, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (342R-14)
vial of 0.1 mL concentrate Research Use Only (342R-14-RUO)
vial of 0.5 mL concentrate (342R-15)
vial of 1.0 mL concentrate (342R-16)
vial of 1.0 mL concentrate Research Use Only (342R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (342R-17-RUO)
vial of 1.0 mL pre-dilute ready-to-use (342R-17)
vial of 7.0 mL pre-dilute ready-to-use (342R-18)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (342R-18-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200 (concentrated)

isotype

IgG

control

uterus

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... TPSAB1(7177)

Related Categories

General description

Tryptases compose a subfamily of proteinases with trypsin-like activity that are mostly stored in mast cell secretory granules and released into the extracellular environment upon mast cell activation. Several biological functions for tryptases have been proposed, including involvement in inflammatory and allergic responses. Mature mast cells have a complex distribution throughout the body. Anti-tryptase is a useful marker for mast cells.

Quality


IVD

IVD

IVD

RUO

Linkage

Tryptase Positive Control Slides, Product No. 342S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Fiorucci et al.
Cellular and molecular life sciences : CMLS, 61(11), 1278-1295 (2004-06-02)
Tryptases constitute a subfamily of trypsin-like proteinases, stored in the mast cell secretory granules of all mammalian organisms. These enzymes are released along with other mediators into the extracellular medium upon mast cell activation/degranulation. Among the trypsin-like enzymes, tryptases are
I S Roberts et al.
Journal of clinical pathology, 53(11), 858-862 (2000-12-29)
Mast cells, when activated, secrete a large number of fibrogenic factors and have been implicated in the development of fibrotic conditions of the liver, lung, and skin. There is evidence that renal fibrosis is closely linked with a chronic inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service