27667
Collagenase Chromophore-Substrate
chromogenic, powder or crystals
Synonym(s):
4-Phenylazobenzyloxycarbonyl-Pro-Leu-Gly-Pro-D-Arg-OH dihydrate
About This Item
Recommended Products
Product Name
Collagenase Chromophore-Substrate, for use at 320 nm
description
for quantitative collagenase determination
form
powder or crystals
shipped in
wet ice
storage temp.
−20°C
SMILES string
O.O.CC(C)C[C@H](NC(=O)[C@@H]1CCCN1C(=O)OCc2ccc(cc2)\N=N\c3ccccc3)C(=O)NCC(=O)N4CCC[C@H]4C(=O)N[C@H](CCCNC(N)=N)C(O)=O
InChI
1S/C38H52N10O8.2H2O/c1-24(2)21-29(33(50)42-22-32(49)47-19-7-12-30(47)34(51)43-28(36(53)54)11-6-18-41-37(39)40)44-35(52)31-13-8-20-48(31)38(55)56-23-25-14-16-27(17-15-25)46-45-26-9-4-3-5-10-26;;/h3-5,9-10,14-17,24,28-31H,6-8,11-13,18-23H2,1-2H3,(H,42,50)(H,43,51)(H,44,52)(H,53,54)(H4,39,40,41);2*1H2/b46-45+;;/t28-,29+,30+,31+;;/m1../s1
InChI key
PZUVBRODNGRBBH-UHGWVMIQSA-N
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service