Skip to Content
Merck
All Photos(1)

Key Documents

Y0000710

Hydrocodone for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hydrocodone bitartrate salt hemi(pentahydrate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO3 · C4H6O6 · 2.5H2O
CAS Number:
Molecular Weight:
494.49
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hydrocodone

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N1([C@H]2[C@H]3[C@@]4([C@@H](Oc5c4c(ccc5OC)C2)C(=O)CC3)CC1)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O.O

InChI

1S/C18H21NO3.C4H6O6.H2O/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19;5-1(3(7)8)2(6)4(9)10;/h3,6,11-12,17H,4-5,7-9H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10);1H2/t11-,12+,17-,18-;1-,2-;/m01./s1

InChI key

ZICRJLQTAUVSDU-NSJZFRMTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydrocodone for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of codeine, morphine, hydrocodone, hydromorphone, oxycodone, and 6-acetylmorphine in urine, serum, plasma, whole blood, and meconium by LC-MS-MS.
Coles R, et al.
Journal of Analytical Toxicology, 31(1), 1-14 (2007)
Forensic drug testing for opiates. VI. Urine testing for hydromorphone, hydrocodone, oxymorphone, and oxycodone with commercial opiate immunoassays and gas chromatography-mass spectrometry.
Smith M L, et al.
Journal of Analytical Toxicology, 19(1), 18-26 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service