Skip to Content
Merck
All Photos(1)

Documents

Y0000361

Ciclosporin for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cyclosporin A, Antibiotic S 7481F1, Cyclosporine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Beilstein:
3647785
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ciclosporin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-CGLBZJNRSA-N

Gene Information

human ... PPIA(5478)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciclosporin for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcel B M Teunissen et al.
The Journal of investigative dermatology, 134(12), 2898-2907 (2014-06-20)
IL-17A is pivotal in the etiology of psoriasis, and CD8(+) T cells with the ability to produce this cytokine (Tc17 cells) are over-represented in psoriatic lesions. Here we demonstrate that the frequency of Tc17 cells in peripheral blood of psoriasis
Yasuo Morishima et al.
Blood, 125(7), 1189-1197 (2014-12-19)
We hypothesized that the compatibility of each HLA loci between donor and patient induced divergent transplant-related immunologic responses, which attributed to the individualized manifestation of clinical outcomes. Here, we analyzed 7898 Japanese pairs transplanted with T-cell-replete marrow from an unrelated
Anne I Dipchand et al.
Pediatric transplantation, 18(7), 764-770 (2014-08-15)
Cardiac remodeling is associated with plasma biomarkers of fibrinogenesis, inflammation, and oxidative stress, and upregulation of mitogenic, pro-fibrotic, and apoptotic signaling pathways. Our primary objective was to evaluate biomarker and subcellular myocardial changes in pediatric heart transplant recipients. Fifty-two-week prospective
Andreas Lossius et al.
European journal of immunology, 44(11), 3439-3452 (2014-08-12)
Epstein-Barr virus (EBV) has long been suggested as a pathogen in multiple sclerosis (MS). Here, we used high-throughput sequencing to determine the diversity, compartmentalization, persistence, and EBV-reactivity of the T-cell receptor (TCR) repertoires in MS. TCR-β genes were sequenced in
David A Williams et al.
Pediatric blood & cancer, 61(5), 869-874 (2013-11-29)
Randomized clinical trials in pediatric aplastic anemia (AA) are rare and data to guide standards of care are scarce. Eighteen pediatric institutions formed the North American Pediatric Aplastic Anemia Consortium to foster collaborative studies in AA. The initial goal of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service