VT000097
Enterococcus faecalis WDCM 00009 Vitroids™
50,000-150,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Certified reference material, Microbiological standards, Enterococcus WDCM 00009 Vitroids™
About This Item
Recommended Products
grade
certified reference material
Quality Level
Agency
ISO 17034
ISO/IEC 17025
form
solid
shelf life
limited shelf life, expiry date on the label
packaging
package of 10 discs
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
application(s)
microbiology
storage temp.
−20°C
suitability
Enterococcus faecalis
Enterococcus spp.
Looking for similar products? Visit Product Comparison Guide
General description
The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms.
These microorganisms are certified according to ISO/IEC 17025 and produced under reproducible conditions compliant with ISO 17034 using authenticated strains from CECT®. These ready-to-use microbiological controls minimize the need for maintaining control strains in the test laboratory and guarantee the use of an authenticated control culture for every quality control test.
{TM="Vitroids"] are derived from a traceable culture obtained freeze-dried from CECT®.
Application
Features and Benefits
- Ready-to-use at convenient CFU ranges
- No dilution required
- Quick rehydration in 10 minutes
- 2-3 years of shelf life in a standard -20 °C freezer
- Complete with a comprehensive certificate of analysis
- No maintenance of stock cultures
- No recovery time or pre-enrichment step is necessary
- Robust, efficient, and reliable
- Easy-to-use
- Cost-effective
Other Notes
Other cell culture collections:
- DSM 20478
- ATCC® 19433
- NCTC 3191
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Interview: Accurate microbiology QC methods for infant formula testing
Interview: Accurate microbiology QC methods for infant formula testing
Interview: Accurate microbiology QC methods for infant formula testing
Interview: Accurate microbiology QC methods for infant formula testing
Related Content
Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.
Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.
Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.
Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service