Skip to Content
Merck
All Photos(1)

Key Documents

SYNPLE-H011

Sigma-Aldrich

Reagent-cartridge Morpholine-2-spiro-(4-Pip)

Synple

Synonym(s):

Synple 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161503
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

parameter

up to 0.5 mmol scale

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C

InChI key

HVPZHRHEHKZOOJ-TYBLODHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reagent-cartridge Morpholine-2-spiro-(4-Pip) is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the synthesis of a morpholine-piperidine spirocycle from an aldehyde through the use of a SnAP reagent. The cartridge contains all necessary reagents for transforming the chosen aldehyde into a morpholine-piperidine spirocycle.

Scale: up to 0.5 mmol
Heterocycle: Morpholine-piperdine spirocycle
Compatible substrates: aldehyde

Cartridge contents:
  • Immobilized SnAP reagent
  • Cu catalyst (Cu(OTF)2 and 2,6-lutidinium triflate)
  • SCX (for purification)
  • Silica (for purification)

Related Cartridges:
  • SYNPLE-H001Reagent-cartridge Morpholine
  • SYNPLE-H002Reagent-cartridge Oxazepane
  • SYNPLE-H004Reagent-cartridge 2-Methylmorpholine
  • SYNPLE-H005Reagent-cartridge Piperazine
  • SYNPLE-H006Reagent-cartridge Diazepane
  • SYNPLE-H007Reagent-cartridge 3-Methylmorpholine
  • SYNPLE-H010Reagent-cartridge Morpholine-2-spiro-(2-Pyr)
  • SYNPLE-H090Reagent-cartridge 9-OMe-Benzoxazepane
  • SYNPLE-H091Reagent-cartridge 7-Br-9-OMe-Benzoxazepane

Learn more about the Synple automated synthesis platform.

Features and Benefits of Synple Platform
  • Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
  • Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety - all chemicals are contained in the system, minimizing the user′s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system - simple installation
  • Reaction temperatures from 25 °C - 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Other Notes

An integrated console for capsule-based, automated organic synthesis

For more information on heterocycle formation using SnAP reagents, visit here: SnAP Technology Spotlight

Legal Information

Product of Synple Chem AG

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service