Skip to Content
Merck

PHR2014

Triamterene Related Compound A

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,4,6-Triamino-5-nitrosopyrimidine, 2,4,6-triamino-5-nitrosopyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H6N6O
CAS Number:
Molecular Weight:
154.13
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to Ph. Eur. N1000000, traceable to USP 1680018

API family

triamterene

CofA

current certificate can be downloaded

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

Nc1nc(c(c(n1)N)N=O)N

InChI

1S/C4H6N6O/c5-2-1(10-11)3(6)9-4(7)8-2/h(H6,5,6,7,8,9)

InChI key

XLQQJSWJHHKLOK-UHFFFAOYSA-N

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

Triamterene Related Compound A may be used as a pharmaceutical reference standard for the determination of impurities in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB5615 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.


Still not finding the right product?

Explore all of our products under Triamterene Related Compound A


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library