Skip to Content
Merck
All Photos(1)

Key Documents

PHR1803

Supelco

Paclitaxel

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Paclitaxel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91
Beilstein:
1420457
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000698
traceable to USP 1491332

API family

paclitaxel

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

213 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](O)[C@@]3(C)C(=O)[C@H](OC(C)=O)C4=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)c5ccccc5)[C@]3([H])[C@@]1(CO2)OC(C)=O)C4(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccccc7

InChI

1S/C47H51NO14/c1-25-31(60-43(56)36(52)35(28-16-10-7-11-17-28)48-41(54)29-18-12-8-13-19-29)23-47(57)40(61-42(55)30-20-14-9-15-21-30)38-45(6,32(51)22-33-46(38,24-58-33)62-27(3)50)39(53)37(59-26(2)49)34(25)44(47,4)5/h7-21,31-33,35-38,40,51-52,57H,22-24H2,1-6H3,(H,48,54)/t31-,32-,33+,35-,36+,37+,38-,40-,45+,46-,47+/m0/s1

InChI key

RCINICONZNJXQF-MZXODVADSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Paclitaxel is a potent anti-neoplastic and anti-mitotic taxane drug, which binds to the N-terminus of β-tubulin and and stabilizes microtubules arresting the cell cycle at the G2/M phase.
Paclitaxel is a potent anti-neoplastic and anti-mitotic taxane drug, which binds to the N-terminus of β-tubulin and and stabilizes microtubules arresting the cell cycle at the G2/M phase. The microtubule damage induces apoptosis through a JNK-dependent pathway followed by a JNK-independent pathway, perhaps related to the activation of protein kinase A (PKA) or of Raf-1 kinase, resulting in phosphorylation of Bcl-2. A major metabolite via CYP2CB is 6α-hydroxypaclitaxel (6α-OHP).

Caution

Paclitaxel undergoes transesterification in methanol and hydrolyzes in aqueous solutions.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9808 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1B - STOT RE 1

Target Organs

Central nervous system,Bone marrow,Cardio-vascular system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erdem D Tabdanov et al.
Cell reports, 25(2), 328-338 (2018-10-12)
Cancer cell migration through and away from tumors is driven in part by migration along aligned extracellular matrix, a process known as contact guidance (CG). To concurrently study the influence of architectural and mechanical regulators of CG sensing, we developed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service