Skip to Content
Merck

PHL80337

Eupatilin

phyproof® Reference Substance

Synonym(s):

2-(3,4-Dimethoxyphenyl)-5,7-dihydroxy-6-methoxy-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3′,4′,6-trimethoxyflavone, NSC 122413

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H16O7
CAS Number:
Molecular Weight:
344.32
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

primary reference standard

product line

phyproof® Reference Substance

assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

SMILES string

O=C1C=C(C2=CC=C(OC)C(OC)=C2)OC3=CC(O)=C(OC)C(O)=C31

InChI

1S/C18H16O7/c1-22-12-5-4-9(6-14(12)23-2)13-7-10(19)16-15(25-13)8-11(20)18(24-3)17(16)21/h4-8,20-21H,1-3H3

InChI key

DRRWBCNQOKKKOL-UHFFFAOYSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Eupatilin is a flavone with anti-oxidative, anti-inflammatory, and anti-cancer properties.
Flavone with anti-oxidative, anti-inflammatory, and anti-cancer properties

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG


Still not finding the right product?

Explore all of our products under Eupatilin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library