Skip to Content
Merck

ERMFD101B

Silica, colloidal

ERM®, certified reference material, 80 nm nominal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

ERM®

manufacturer/tradename

JRC

application(s)

general analytical

Legal Information

ERM is a registered trademark of European Commission


Still not finding the right product?

Explore all of our products under Silica, colloidal


Storage Class

12 - Non Combustible Liquids

wgk

nwg



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library