Skip to Content
Merck
All Photos(1)

Key Documents

BCR136R

Benzo[b]naphtho[2,3-d]thiophene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10S
CAS Number:
Molecular Weight:
234.32
Beilstein:
157102
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2cc3c(cc2c1)sc4ccccc34

InChI

1S/C16H10S/c1-2-6-12-10-16-14(9-11(12)5-1)13-7-3-4-8-15(13)17-16/h1-10H

InChI key

UWMISBRPSJFHIR-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR136R

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Jacob et al.
Cancer letters, 32(1), 107-116 (1986-07-01)
Thiaarenes are metabolized by liver microsomes of untreated rats predominantly to sulfones and sulfoxides. After pretreatment of rats with monooxygenase inducers, ring oxidation of thiaarenes is also observed. In case of benzo[b]naphtho[2,3-d]thiophene the formation of a p-quinone takes place. Rat
Hannie S van der Honing et al.
The New phytologist, 185(1), 90-102 (2009-09-19)
Here, we produced cytoplasmic protrusions with optical tweezers in mature BY-2 suspension cultured cells to study the parameters involved in the movement of actin filaments during changes in cytoplasmic organization and to determine whether stiffness is an actin-related property of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service