Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3(CH2)13CO2CH3
CAS Number:
Molecular Weight:
256.42
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
230-430-6
Beilstein/REAXYS Number:
1776383
MDL number:
grade
analytical standard
Quality Segment
assay
≥99.5% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
refractive index
n20/D 1.438 (lit.), n20/D 1.439
bp
141-142 °C/3 mmHg (lit.)
mp
18-19 °C (lit.)
density
0.865 g/mL at 20 °C, 0.862 g/mL at 25 °C (lit.)
format
neat
functional group
ester
shipped in
ambient
storage temp.
room temp
SMILES string
CCCCCCCCCCCCCCC(=O)OC
InChI
1S/C16H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18-2/h3-15H2,1-2H3
InChI key
XIUXKAZJZFLLDQ-UHFFFAOYSA-N
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Methyl pentadecanoate may be used as:
- A standard for the quantitation of monoepoxy fatty acid methyl esters (FAME) by gas-liquid chromatography.
- An internal standard for the extraction of triglycerides from de-shelled Jatropha curcas L. seeds by use of supercritical carbon dioxide (SC-CO2) extraction.
Still not finding the right product?
Explore all of our products under Methyl pentadecanoate
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
