70289
Sodium 2-naphthalenesulfonate
suitable for ion pair chromatography, LiChropur™, ≥99.0% (HPLC)
Synonym(s):
2-Naphthalenesulfonic acid sodium salt
About This Item
Recommended Products
description
anionic
Quality Level
Assay
≥99.0% (HPLC)
form
powder
quality
LiChropur™
technique(s)
ion pair chromatography: suitable
λ
10 % in H2O (hot)
UV absorption
λ: 380 nm Amax: 0.1
λ: 390 nm Amax: 0.05
λ: 400 nm Amax: 0.02
λ: 500 nm Amax: 0.02
suitability
corresponds to standard for filter test
SMILES string
[Na+].[O-]S(=O)(=O)c1ccc2ccccc2c1
InChI
1S/C10H8O3S.Na/c11-14(12,13)10-6-5-8-3-1-2-4-9(8)7-10;/h1-7H,(H,11,12,13);/q;+1/p-1
InChI key
YWPOLRBWRRKLMW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service