Skip to Content
Merck
All Photos(1)

Documents

67425

Millipore

MDR Acinetobacter Millichrome plus Supplement

selective for bacteria: Acinetobacter spp. (MDR), suitable for microbiology

Synonym(s):

MDR Acinetobacter Growth Media plus Supplement, MDR Acinetobacter Chromogenic Medium plus Supplement, MDR Acinetobacter Culture Media plus Supplement, MDR Acinetobacter Millichrome+ Supplement, MDR Acinetobacter Selective Supplement, Multi-Drug Resistant Acinetobacter Millichrome plus Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

sterility

sterile; aseptically filled

Quality Level

form

dry powder

shelf life

limited shelf life, expiry date on the label

technique(s)

microbiological culture: suitable

solubility

sterile water: soluble (1 vial per 5ml)

application(s)

microbiology
pharmaceutical
water monitoring

shipped in

ambient

storage temp.

2-8°C

suitability

selective for bacteria: Acinetobacter spp. (MDR)

General description

The MDR Acinetobacter Millichrome plus Supplement is a selective supplement is high selectivity for multi-drug resistant Acinetobacter strains. The package contains 5 vials, with each vial sufficient for 1 liter of final media. One vial of the supplement should be rehydrated with 5 mL of purified water and then added to 1 liter of melted Acinetobacter Millichrome plus Agar base at 45-50 °C.

Application

MDR Acinetobacter Millichrome plus Supplement is a selective supplement used for screening multi-drug resistant Acinetobacter to aid in the prevention and control of multi-drug resistant (MDR) Acinetobacter, in healthcare settings and pharma production. It can also be used in hygiene monitoring in the clinical and pharma environment with surface sampling.

Packaging

5 vials (1 vial is sufficient for 1 L of final media)

Preparation Note

  • Rehydrate one vial with 5 mL of purified water.
  • Add 5 mL of this solution to 1 L of the melted Acinetobacter Millichrome® plus Agar base at 45-50 °C.
  • Stir well for homogenization

Storage and Stability

Store the lyophilized powder, in a dry place, in tightly sealed containers at 2-8°C.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service