Skip to Content
Merck
All Photos(2)

Key Documents

37038

Supelco

Flumorph

PESTANAL®, analytical standard

Synonym(s):

(EZ)-4-[3-(3,4-Dimethoxyphenyl)-3-(4-fluorophenyl)acryloyl]morpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22FNO4
CAS Number:
Molecular Weight:
371.40
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1ccc(cc1OC)\C(=C/C(=O)N2CCOCC2)c3ccc(F)cc3

InChI

1S/C21H22FNO4/c1-25-19-8-5-16(13-20(19)26-2)18(15-3-6-17(22)7-4-15)14-21(24)23-9-11-27-12-10-23/h3-8,13-14H,9-12H2,1-2H3/b18-14+

InChI key

BKBSMMUEEAWFRX-NBVRZTHBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flumorph is a oomycete carboxylic acid amide (CAA) fungicide, which shows strong activity against Peronospora and Phytophthora diseases in vegetables and fruits.

Application

Flumorph may be used as an analytical reference standard for the quantification of the analyte in vegetables, fruits and biological samples using different chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of flumorph residues in vegetables, soil, and natural water by solid-phase extraction cleanup and high-performance liquid chromatography with UV detection
Hu Y-J, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 91(6), 1459-1466 (2008)
A sensitive and selective method for the determination of flumorph residues in vegetables and fruits by HPLC-ESI-MS/MS
Wang Y, et al.
Talanta, 86, 221-226 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service