Skip to Content
Merck

32772

Aflatoxin G1-13C17 solution

~0.5 μg/mL in acetonitrile, analytical standard

Synonym(s):

Fully13C-labelled Aflatoxin G1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
13C17H12O7
CAS Number:
Molecular Weight:
345.15
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
EC Number:
200-835-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

concentration

~0.5 μg/mL in acetonitrile

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cannabis testing
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+17

storage temp.

−20°C

SMILES string

[13CH3]O[13c]1[13cH][13c]2O[13C@H]3O[13CH]=[13CH][13C@H]3[13c]2[13c]4O[13C](=O)[13C]5=[13C]([13CH2][13CH2]O[13C]5=O)[13c]14

InChI

1S/C17H12O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h3,5-6,8,17H,2,4H2,1H3/t8-,17+/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1

InChI key

XWIYFDMXXLINPU-DENWKYDXSA-N

General description

Aflatoxin G1-13C17 is the 13C isotope-labeled analog of the mycotoxin, aflatoxin.
Certan Vial

Application

Aflatoxin G1-13C17 may be used as an analytical standard for the determination of aflatoxin G1 in food grade gums as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

35.6 °F - closed cup

Flash Point (°C)

2.0 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library