Skip to Content
Merck

00291

Chlormequat-1,1,2,2-d4 chloride

PESTANAL®, analytical standard

Synonym(s):

(2-Chloroethyl-d4)trimethylammonium chloride, Chlorocholine-1,1,2,2-d4 chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5D4H9Cl2N
Molecular Weight:
162.09
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
77101502
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

isotopic purity

≥99 atom % D

product line

PESTANAL®

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

SMILES string

ClC([2H])([2H])C([2H])([2H])[N+](C)(C)C.[Cl-]

InChI

1S/C5H13ClN.ClH/c1-7(2,3)5-4-6;/h4-5H2,1-3H3;1H/q+1;/p-1/i4D2,5D2;

InChI key

UHZZMRAGKVHANO-HGFPCDIYSA-M

Application


  • Surface-enhanced Raman spectroscopy charged probes under inverted superhydrophobic platform for detection of agricultural chemicals residues in rice combined with lightweight deep learning network: This study demonstrates a novel application of Chlormequat-1,1,2,2-d4 Cl in the detection of agricultural chemicals in rice. The use of surface-enhanced Raman spectroscopy provides enhanced sensitivity and specificity, facilitated by an inverted superhydrophobic platform and integrated with a lightweight deep learning network for efficient analysis (Weng et al., 2023).

  • Highly water-dispersible hydroxyl functionalized covalent organic frameworks as matrix for enhanced MALDI-TOF MS identification and quantification of quaternary ammonium salts in water and fruits: This research explores the use of Chlormequat-1,1,2,2-d4 Cl in enhancing the performance of covalent organic frameworks used as matrices in MALDI-TOF MS for the detection and quantification of quaternary ammonium salts. The hydroxyl functionalization contributes to the dispersibility in water, offering significant improvements in the identification process in water and fruit samples (Zheng et al., 2022).

  • The disrupting effect of chlormequat chloride on growth hormone is associated with pregnancy: This study investigates the biological impact of Chlormequat-1,1,2,2-d4 Cl, specifically its disruptive effects on growth hormone regulation during pregnancy. The findings are critical for understanding the biochemical pathways affected by this compound and its implications in reproductive health (Wu et al., 2024).

  • Chlormequat chloride induced activation of calmodulin mediated PI3K/AKT signaling pathway led to impaired sperm quality in pubertal mice: This publication details the detrimental effects of Chlormequat-1,1,2,2-d4 Cl on sperm quality in mice, mediated through the activation of the calmodulin-dependent PI3K/AKT signaling pathway. Such studies are vital for evaluating the reproductive toxicology of Chlormequat and related compounds (Wang et al., 2024).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

No data available

flash_point_c

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library