Skip to Content
Merck
All Photos(1)

Key Documents

ZFPACKSP0

Milli-Q

Pack Support 2 Tube

without gauges, For secure, leak-free fit of the pretreatment pack

Synonym(s):

Pack Support 2 Tube without guages

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
NACRES:
JA.13

packaging

pkg of 1 unit

compatibility

for use with Direct-Q® UV
for use with Direct-Q®

Looking for similar products? Visit Product Comparison Guide

General description

The Pack Support 2 tube is a water purification accessory that provides a leak-free fit of pretreatment pack.

Application

Wall mounting bracket for installation and operation of pretreatment pack

Legal Information

DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service