Skip to Content
Merck

SCR545

Human STEMCCA Cre-Excisable Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12161503
NACRES:
NA.81
eCl@ss:
32161000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Induced Pluripotent Stem Cells

Quality Segment

technique(s)

cell culture | stem cell: suitable

General description

The Human STEMCCA Cre-Excisable Constitutive Polycistronic (OKSM) Lentivirus Kit contains high titer polycistronic (OKSM) lentivirus and Polybrene transfection reagent that have been validated for the generation of human induced pluripotent stem cells from human foreskin fibroblasts (HFFs). The Cre-excisable STEMCCA vector, comprised of the transcription factors human Oct-4, Klf4, SOX-2, and c-Myc (OKSM) separated by the self-cleaving 2A peptide and IRES sequences, includes flanking LoxP sites that enable Cre-mediated excision of the viral transgenes following reprogramming (Sommer CA, 2009).

The use of a single polycistronic lentiviral vector significantly improves reprogramming efficiencies and reduces the number of viral integrations. Furthermore, the ability to remove the reprogramming vector has been shown to improve the developmental potential of iPS cells and significantly increase their capacity to undergo directed differentiation in vitro (Sommer CA, 2010).

Preparation Note

Lentivirus is stable for at least 6 months when stored at -80°C. After first thaw, place immediately on ice and store in working aliquots to avoid further freeze thaws. Avoid freeze thaws as this will result in a decrease in the virus titer. Polybrene transfection reagent is stable for at least 1 year when stored at -20°C.

Analysis Note

Tested to confirm the generation of iPS cells from p6 human foreskin fibroblasts. Other cell types have not been tested and thus similar results can not be guaranteed.

Other Notes

1. Human STEMCCA Cre-Excisable Constitutive (OKSM) Lentivirus: (Part number SCR545-1) Two (2) vials of EF1α-hSTEMCCA-LoxP (OKSM) Lentivirus (Part number CS204503). Each vial contains 15 µL of high titer lentivirus. 2. Polybrene Transfection Reagent: (Part number TR-1003-50UL) One (1) vial containing 50 µL of 10 mg/mL stock.

Legal Information

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library