Skip to Content

Legal compliance reminder: purchasers must obtain all necessary licenses and comply with applicable laws (see General Terms & Conditions of Sale). We reserve the right to request additional information.

Merck

Skip To

SCC295

hERG1a/1b Inducible HEK293 Cell Line

Human

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106514
NACRES:
NA.71
Biological source:
human
Growth mode:
N/A
Manufacturer/tradename:
Sigma-Aldrich
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

hERG1a/1b Inducible HEK293 Cell Line,

biological source

human

packaging

pkg of >1X10⁶ vials

manufacturer/tradename

Sigma-Aldrich

growth mode

N/A

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

≤ − 140°C

General description

HERG channel inhibition assays for preclinical cardiac safety screening of new drug compounds use electorphysiological methodology and employ hERG-expressing recombinant cell lines. Despite evidence that the cardiac IKr potassium current is produced by heteromeric hERG1a/1b channels1, 2, 3, drug screening assays have long relied on homomeric hERG1a alone, because of the instability of hERG 1b expression in so-called “stable” recombinant cell lines4. The novel inducible hERG1a/1b heteromeric cell line offers an improved tool for preclinical cardiac toxicity screening, by enabling more accurate predictions of off-target drug effects on cardiac IKr, particularly in computational models relying on accurate IC50 measurements. hERG1a/1b cells also can facilitate the study of heteromeric assembly of the hERG 1a and 1b subunits during biogenesis.5

Application

HERG channel inhibition assays for preclinical cardiac safety screening of new drug compounds use electorphysiological methodology and employ hERG-expressing recombinant cell lines. Despite evidence that the cardiac IKr potassium current is produced by heteromeric hERG1a/1b channels1, 2, 3, drug screening assays have long relied on homomeric hERG1a alone, because of the instability of hERG 1b expression in so-called “stable” recombinant cell lines4. The novel inducible hERG1a/1b heteromeric cell line offers an improved tool for preclinical cardiac toxicity screening, by enabling more accurate predictions of off-target drug effects on cardiac IKr, particularly in computational models relying on accurate IC50 measurements. hERG1a/1b cells also can facilitate the study of heteromeric assembly of the hERG 1a and 1b subunits during biogenesis.5 Source The heteromeric hERG1a/1b Inducible HEK293 cell line, in which hERG1b protein expression is tightly controlled by doxycyclin dose and treatment time, was derived from a homomeric hERG 1a expressing HEK293 cell line.5 References1. J Biol Chem 2004, 279(43): 44690–446942. Circ Res 2008, 103(7): e81–e953. PNAS 2014, 111(50): 18073-180774. Br J Pharmacol 2011, 164(2b): 419-432. 5. Biophysical J 2018, Volume 114, Issue 3, Suppl 1, 2 (292a) (Poster Abstract)
• Each vial contains ≥ 1X10⁶ viable cells.• Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.• Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services• Cells are negative for mycoplasma contamination.

Features and Benefits

The heteromeric hERG1a/1b Inducible HEK293 cell line, in which hERG1b protein expression is tightly controlled by doxycyclin dose and treatment time, was derived from a homomeric hERG 1a expressing HEK293 cell line

Preparation Note

The cells should be stored in liquid nitrogen until use. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting functionality.

Other Notes

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact licensing@milliporesigma.com.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCC461SCC291SCC109
description

-

description

-

description

-

description

AC16 Human Cardiomyocytes can be serially passaged and can differentiate when cultured in mitogen-free medium. The cells may be used to study developmental regulation of cardiomyocytes.

growth mode

N/A

growth mode

N/A

growth mode

N/A

growth mode

-

biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable, receptor binding assay: suitable, cell based assay: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

≤ − 140°C

storage temp.

−196°C

storage temp.

≤ − 140°C

storage temp.

-140 to -196°C

shipped in

dry ice

shipped in

liquid nitrogen

shipped in

dry ice

shipped in

dry ice

manufacturer/tradename

Sigma-Aldrich

manufacturer/tradename

Millipore

manufacturer/tradename

Sigma-Aldrich

manufacturer/tradename

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters