Skip to Content
Merck
All Photos(1)

Key Documents

PX1476

Supelco

Potassium Hydrogen Phthalate

Meets ACS Specifications GR ACS

Synonym(s):

Potassium Hydrogen Phthalate, Potassium Biphthalate, Potassium Acid Phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5KO4
CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
41116107
EC Index Number:
212-889-4
Assay:
99.95-100.05% dry basis
form:
solid
solubility:
80 g/L

Quality Level

Assay

99.95-100.05% dry basis

form

solid

impurities

≤0.002% Sulfur compounds (as S)
≤0.005% Insoluble matter

color

white

pH

4.0 (20 °C, 50 g/L in H2O)

mp

295-300 °C

solubility

80 g/L

anion traces

chloride (Cl-): 0.003%

cation traces

Fe: ≤5 ppm
Na: ≤0.005%
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[K+].[O-]C(=O)c1c(cccc1)C(=O)[O-].[H+]

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Related Categories

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay (dried basis)99.95-100.05%Chlorine compounds (as Cl)0.003% maxColorWhiteFormCrystalsHeavy metals (as Pb)5ppm maxInsoluble matter0.005% maxIron (Fe)5ppm maxSodium (Na)0.005% maxSulfur compounds (as S)0.002% maxpH of a 0.05 m solution at 25.0 ± 0.2°C4.00-4.02

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service