Skip to Content
Merck
All Photos(4)

Documents

PFHYS0616

Millipore

Millicell® HY Multilayer Culture Flask

T-600, 3 layer, 120-180 mL, sterile, vented cap, Tissue Culture (TC)-treated surface

Synonym(s):

High Yield Culture Flask, Millicell HY Flask T-600 Cell Culture Flask, cell culture, cell culture flask, multi-layer flask, tissue culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22

material

angled neck high-density polyethylene cap (vented)
polystyrene flask

Quality Level

sterility

sterile; γ-irradiated

feature

3 layer

packaging

pack of 16

manufacturer/tradename

Millicell®

parameter

45 °C max. temp.

technique(s)

cell culture | mammalian: suitable

H

32 mm , excluding cap
51 mm , including cap

L

22.2 cm

W

12 cm

surface area

600 cm2

working volume

120-180 mL

binding type

Tissue Culture (TC)-treated surface

shipped in

ambient

Preparation Note

  1. To seed cells, fill the Millicell HY T-600 Flask with the desired growth media.
  2. Add desired number of cells to achieve typical seeding density.
  3. Mix the cells and media by raising the end of the flask and pipetting the solution up and down several times until a homogeneous suspension is achieved. Note: Failure to adequately mix cells into a homogeneous suspension may result in an uneven distribution of cells across the layers of the Millicell HY Flask.
  4. Briefly lay the flask on its side to equilibrate the liquid volume equally among the layers of the flask.
  5. Tilt the flask on its end to partition the liquid volume for each layer of the flask.
  6. Lay the flask down to spread the cell suspension evenly across the entire surface of each layer.
  7. Depending on the volume it may be necessary to rock the flask on all four corners to ensure that the entire surface wets. Note: If the liquid volume on any of the layers changes during the wetting procedure (e.g., spilling of liquid from top layer to lower layers), repeat steps 4–6 to ensure even distribution of liquid among all layers.
  8. Return the flask to a vertical position when transporting to the incubator. Lay the flask flat in the incubator (as in step 6) and culture cells under appropriate conditions.
  9. If media exchange is required, either aspirate or pour the media from the Millicell HY T-600 Flask. Note: Due to the larger surface area of the Millicell Flasks, it may be necessary to let the flask stand on its side for several minutes following each aspiration so that all residual liquid pools to a common point, allowing for a second aspiration to completely remove the remaining liquid.
  10. Add appropriate volume of fresh media, and repeat steps 4–8.
  11. To harvest cells, remove media as in step 9.
  12. Add desired amount of appropriate wash solution (e.g., PBS or 0.02% EDTA) to the flask and repeat steps 4–7.
  13. Remove wash solution as in step 9.
  14. Add desired amount of dissociation enzyme (e.g., 0.25% trypsin/EDTA or equivalent) according to preferred protocol, repeat steps 4–8, and incubate. Volume of dissociation enzyme required per cm2 does not need to be altered from T-flask protocol. Note: Ensure that all cells have completely dissociated from the surface of the flask via observation under microscope. Failure to allow complete dissociation of cells from culture surface (such that all cells are freely floating) may result in reduced cell yields.
  15. If using trypsin, add desired volume of inactivating solution, such as serum-containing media.
  16. Collect cells either by aspirating or pouring, as in step 9.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Monitor your cell cultures with ease with the Millicell® DCI Digital Cell Imager. From confluency to cell count and morphology, the Millicell® DCI automates cell culture measurements for quicker, more consistent culturing.

Monitor your cell cultures with ease with the Millicell® DCI Digital Cell Imager. From confluency to cell count and morphology, the Millicell® DCI automates cell culture measurements for quicker, more consistent culturing.

Monitor your cell cultures with ease with the Millicell® DCI Digital Cell Imager. From confluency to cell count and morphology, the Millicell® DCI automates cell culture measurements for quicker, more consistent culturing.

Monitor your cell cultures with ease with the Millicell® DCI Digital Cell Imager. From confluency to cell count and morphology, the Millicell® DCI automates cell culture measurements for quicker, more consistent culturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service