Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
packaging
pkg of 1 pack
manufacturer/tradename
Millipore
technique(s)
SDS-PAGE: suitable, electrophoresis: suitable
compatibility
for use with mPAGE® Mini Wet Transfer System
General description
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description replacement part for mPAGE® Mini Wet Transfer System | description replacement part for mPAGE® Mini Wet Transfer System | description for transfer of proteins from a mini acrylamide gel to blotting membrane, accommodates two wet transfer cassettes | description for electrophoresis of precast gels or hand cast polyacrylamide gels |
| compatibility for use with mPAGE® Mini Wet Transfer System | compatibility for use with mPAGE® Mini Wet Transfer System. | compatibility for use with mPAGE® Mini Gel Tank | compatibility for use with mPAGE®
|
| packaging pkg of 1 pack | packaging pkg of 1 pack | packaging pkg of 1 system, pkg of 1 ea ( for transferring up to 2 gels) | packaging 1 ea of (for running 2 or 4 gels) |
| manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore |
| technique(s) SDS-PAGE: suitable | technique(s) SDS-PAGE: suitable, electrophoresis: suitable | technique(s) immunoblotting: suitable, western blot: suitable | technique(s) SDS-PAGE: suitable, electrophoresis: suitable |
Choose from one of the most recent versions:
Already Own This Product?