Skip to Content
Merck

MSP010063

Petri-Pad Petri dish EO sterilized

Petridish with Petri-Pad for m-ColiBlue 24 broth application

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122101
NACRES:
NB.14
eCl@ss:
32030209
Material:
cellulose pad(s), clear device, polystyrene
Sterility:
(Sterilization; ethylene oxide treated)
Diam.:
47 mm
Packaging:
pkg of 150 ×
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

cellulose pad(s), clear device, polystyrene

sterility

(Sterilization; ethylene oxide treated)

packaging

pkg of 150 ×

diam.

47 mm

input

sample type beer
sample type soft drink
sample type bottled water
sample type dairy
sample type wine
sample(s) (environmental)
sample type beverage(s)
sample type cider
sample type food(s)

application(s)

bioburden testing
environmental
environmental (monitoring)
food and beverages

shipped in

ambient

General description

Petri-Pad Petri dish is 47mm sterilized Petri dishes for use with solid agar medium. Pre-sterilized disposable 47mm dishes preloaded with sterilized absorbent pads eliminate labor cost of hand-loading pads and minimize contamination. m-ColiBlue24 Broth allows for the simultaneous detection of total coliform bacteria and Escherichia coli (E. coli) within 24 hours. An enzymatic indicator in the medium causes non-fecal total coliform colonies grown on the m-ColiBlue24 medium to be red, while the E. coli (fecal coliform) colonies are blue. The selectivity of the enzymatic indicator eliminates the need for confirmation.

Application

Petridish with Petri-Pad for m-ColiBlue24 medium and is used in detection of total coliform bacteria in Beer, Bottled Water, Cider, Sport Drink, Wine etc.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library